Learn More
Meso Scale Discovery R-PLEX Human cIAP2 ASYQP
SDP
Catalog No. 5000078842
Encompass
The R-PLEX Human CIAP2 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the levels of cellular inhibitor of apoptosis protein 2 (CIAP2) in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls This assay allows for the quantitative measurement of CIAP2 which plays a role in inhibiting apoptosis and promoting cell survival
Catalog No. 50-000-78842 Supplier Meso Scale Discovery Supplier No. K1517WR22
The R-PLEX Human CIAP2 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the levels of cellular inhibitor of apoptosis protein 2 (CIAP2) in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls This assay allows for the quantitative measurement of CIAP2 which plays a role in inhibiting apoptosis and promoting cell survival