Learn More
Meso Scale Discovery R-PLEX Human CKBB ASY
SDP
Catalog No. 5000080624
Encompass
The R-PLEX Human CK-BB Assay Kit from Meso Scale Diagnostics measures creatine kinase isoenzyme BB (CK-BB) in human serum plasma and other fluids using electrochemiluminescence (ECL) detection It offers high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls diluents buffers and seals Sensitivity is 0 3 pg/mL with a quantification limit of 10 000 pg/mL enabling CK-BB measurement a key biomarker for myocardial infarction and cardiovascular conditions
Catalog No. 50-000-80624 Supplier Meso Scale Discovery Supplier No. K1516PR2
The R-PLEX Human CK-BB Assay Kit from Meso Scale Diagnostics measures creatine kinase isoenzyme BB (CK-BB) in human serum plasma and other fluids using electrochemiluminescence (ECL) detection It offers high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls diluents buffers and seals Sensitivity is 0 3 pg/mL with a quantification limit of 10 000 pg/mL enabling CK-BB measurement a key biomarker for myocardial infarction and cardiovascular conditions