Learn More
Meso Scale Discovery R-PLEX Human CNTF ASY
SDP
Catalog No. 5000082433
Encompass
The R-PLEX Human CNTF Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Ciliary Neurotrophic Factor (CNTF) in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents calibrator and controls The assay has a typical lower limit of detection of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL for CNTF
Catalog No. 50-000-82433 Supplier Meso Scale Discovery Supplier No. K151AMAR2
The R-PLEX Human CNTF Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Ciliary Neurotrophic Factor (CNTF) in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents calibrator and controls The assay has a typical lower limit of detection of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL for CNTF