Learn More
Meso Scale Discovery R-PLEX Human CNTF ASYQP
SDP
Catalog No. 5000078498
Encompass
The R-PLEX Human CNTF Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring Ciliary Neurotrophic Factor (CNTF) in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume requirements The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay quantifies CNTF important for neuronal cell survival and differentiation
Catalog No. 50-000-78498 Supplier Meso Scale Discovery Supplier No. K151AMAR22
The R-PLEX Human CNTF Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring Ciliary Neurotrophic Factor (CNTF) in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume requirements The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay quantifies CNTF important for neuronal cell survival and differentiation