Learn More
Meso Scale Discovery R-PLEX Human COMP/TSP-5 ASY
SDP
Catalog No. 5000080368
Encompass
The R-PLEX Human Comp TSP-5 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of 5 different human Th17-related proteins (IL-17A IL-17F IL-21 IL-22 IL-23) in serum plasma cell culture supernatants and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 96-well plate format The kit includes all necessary reagents and diluents needed to perform the assay The sensitive and high-throughput assay allows for the accurate quantification of Th17-related cytokines in approximately 4 hours
Catalog No. 50-000-80368 Supplier Meso Scale Discovery Supplier No. K151U0R2
The R-PLEX Human Comp TSP-5 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of 5 different human Th17-related proteins (IL-17A IL-17F IL-21 IL-22 IL-23) in serum plasma cell culture supernatants and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 96-well plate format The kit includes all necessary reagents and diluents needed to perform the assay The sensitive and high-throughput assay allows for the accurate quantification of Th17-related cytokines in approximately 4 hours