Learn More
Meso Scale Discovery R-PLEX Human Cystatin B ASYQP
SDP
Catalog No. 5000079585
Encompass
The R-PLEX Human Cystatin B Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring Cystatin B in human serum plasma and other fluids It uses MSD s electrochemiluminescent detection technology offering high sensitivity a broad dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies a detection antibody conjugate calibrator solutions and necessary reagents It detects Cystatin B from 19 pg/mL to 100 000 pg/mL with high specificity
Catalog No. 50-000-79585 Supplier Meso Scale Discovery Supplier No. K151AAKR22
The R-PLEX Human Cystatin B Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring Cystatin B in human serum plasma and other fluids It uses MSD s electrochemiluminescent detection technology offering high sensitivity a broad dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies a detection antibody conjugate calibrator solutions and necessary reagents It detects Cystatin B from 19 pg/mL to 100 000 pg/mL with high specificity