Learn More
Meso Scale Discovery R-PLEX Human Delta-like 1 ASY
SDP
Catalog No. 5000079073
Encompass
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a sensitive immunoassay for measuring human interleukin 6 (IL-6) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) technology on the MSD platform for high sensitivity and a broad dynamic range The kit includes a plate with capture antibody detection antibody calibrator and assay diluent Sensitivity is 0 10 pg/mL with an upper quantification limit of 1500 pg/mL The assay time is about 3 5 hours for 96 tests offering a robust tool for studying IL-6 biology and evaluating IL-6-targeted therapeutics
Catalog No. 50-000-79073 Supplier Meso Scale Discovery Supplier No. K151AALR2
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a sensitive immunoassay for measuring human interleukin 6 (IL-6) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) technology on the MSD platform for high sensitivity and a broad dynamic range The kit includes a plate with capture antibody detection antibody calibrator and assay diluent Sensitivity is 0 10 pg/mL with an upper quantification limit of 1500 pg/mL The assay time is about 3 5 hours for 96 tests offering a robust tool for studying IL-6 biology and evaluating IL-6-targeted therapeutics