Learn More
Meso Scale Discovery R-PLEX Human E-Cadherin ASY
SDP
Catalog No. 5000079634
Encompass
The R-Plex Human E-Cadherin Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures E-Cadherin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the quantitative measurement of E-Cadherin an important biomarker that plays a role in cell adhesion and is often monitored in cancer research With minimal hands-on time this robust and reproducible assay generates rapid results using automated instrumentation
Catalog No. 50-000-79634 Supplier Meso Scale Discovery Supplier No. K151YXR2
The R-Plex Human E-Cadherin Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures E-Cadherin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the quantitative measurement of E-Cadherin an important biomarker that plays a role in cell adhesion and is often monitored in cancer research With minimal hands-on time this robust and reproducible assay generates rapid results using automated instrumentation