Learn More
Meso Scale Discovery R-PLEX Human EGFR ASY
SDP
Catalog No. 5000082265
Encompass
The R-PLEX Human EGFR Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay measuring total EGFR levels in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) assay uses MSD s SULFO-TAG labels and multi-array plate technology for simultaneous detection of multiple protein biomarkers from a single sample It offers high sensitivity broad dynamic range low sample volume and rapid results The kit includes a 96-well plate with capture antibody detection antibody calibrator and assay diluent suitable for biomarker discovery disease research and drug development
Catalog No. 50-000-82265 Supplier Meso Scale Discovery Supplier No. K151N5R2
The R-PLEX Human EGFR Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay measuring total EGFR levels in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) assay uses MSD s SULFO-TAG labels and multi-array plate technology for simultaneous detection of multiple protein biomarkers from a single sample It offers high sensitivity broad dynamic range low sample volume and rapid results The kit includes a 96-well plate with capture antibody detection antibody calibrator and assay diluent suitable for biomarker discovery disease research and drug development