Learn More
Meso Scale Discovery R-PLEX Human EGFR ASYQP
SDP
Catalog No. 5000079921
Encompass
The R-Plex Human EGFR Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human epidermal growth factor receptor (EGFR) in cell and tissue lysates The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents and calibrator The assay has a typical sensitivity of 0 3 ng/mL and range up to 2500 ng/mL
Catalog No. 50-000-79921 Supplier Meso Scale Discovery Supplier No. K151N5R22
Add to Cart
Add to Cart
The R-Plex Human EGFR Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human epidermal growth factor receptor (EGFR) in cell and tissue lysates The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents and calibrator The assay has a typical sensitivity of 0 3 ng/mL and range up to 2500 ng/mL