Learn More
Meso Scale Discovery R-PLEX Human Elastase-2 ASY
SDP
Catalog No. 5000080322
Encompass
The R-PLEX Human Elastase 2 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures human elastase 2 levels in serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay is provided in a convenient all-in-one kit including plates coated with capture antibody detection antibody read buffer calibrator and controls The R-PLEX Human Elastase 2 Assay allows for the quantitative measurement of elastase 2 an enzyme involved in tissue remodeling and inflammation
Catalog No. 50-000-80322 Supplier Meso Scale Discovery Supplier No. K151ABKR2
The R-PLEX Human Elastase 2 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures human elastase 2 levels in serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay is provided in a convenient all-in-one kit including plates coated with capture antibody detection antibody read buffer calibrator and controls The R-PLEX Human Elastase 2 Assay allows for the quantitative measurement of elastase 2 an enzyme involved in tissue remodeling and inflammation