Learn More
Meso Scale Discovery R-PLEX Human Endoglin ASY
SDP
Supplier:  Meso Scale Discovery K1510DR2
Encompass
The R-PLEX Human Endoglin Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring endoglin in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators and necessary reagents It measures endoglin in a 7-plex format allowing simultaneous analysis of 6 other analytes useful for studying angiogenesis cardiovascular diseases cancer and related conditions
Catalog No. 50-000-78891