Learn More
Meso Scale Discovery R-PLEX Human Endoglin ASYQP
SDP
Catalog No. 5000078844
Encompass
The R-PLEX Human Endoglin Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring endoglin in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators and necessary reagents It ensures high specificity and minimal cross-reactivity allowing rapid and accurate quantification of endoglin a key biomarker from small samples
Catalog No. 50-000-78844 Supplier Meso Scale Discovery Supplier No. K1510DR22
The R-PLEX Human Endoglin Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring endoglin in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators and necessary reagents It ensures high specificity and minimal cross-reactivity allowing rapid and accurate quantification of endoglin a key biomarker from small samples