Learn More
Meso Scale Discovery R-PLEX Human Endostatin ASY
SDP
Catalog No. 5000078579
Encompass
The Meso Scale Discovery R-PLEX Human Endostatin Assay is a multiplex immunoassay panel that allows simultaneous quantification of endostatin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This assay requires 50 L of sample and can perform up to 80 tests across two plates It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and calibrator containing recombinant proteins This assay allows sensitive reproducible and rapid quantification of endostatin
Catalog No. 50-000-78579 Supplier Meso Scale Discovery Supplier No. K151P8R2
The Meso Scale Discovery R-PLEX Human Endostatin Assay is a multiplex immunoassay panel that allows simultaneous quantification of endostatin in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This assay requires 50 L of sample and can perform up to 80 tests across two plates It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and calibrator containing recombinant proteins This assay allows sensitive reproducible and rapid quantification of endostatin