Learn More
Meso Scale Discovery R-PLEX Human Endostatin ASYQP
SDP
Catalog No. 5000078764
Encompass
The R-PLEX Human Endostatin Assay from Meso Scale Discovery is a multiplex immunoassay that measures endostatin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This assay allows for the quantitative measurement of endostatin an endogenous inhibitor of angiogenesis It requires 50 L of sample and can perform up to 80 tests across two plates The total assay time is 3 5 hours This product could be useful for studying angiogenesis regulation and cancer progression
Catalog No. 50-000-78764 Supplier Meso Scale Discovery Supplier No. K151P8R22
The R-PLEX Human Endostatin Assay from Meso Scale Discovery is a multiplex immunoassay that measures endostatin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This assay allows for the quantitative measurement of endostatin an endogenous inhibitor of angiogenesis It requires 50 L of sample and can perform up to 80 tests across two plates The total assay time is 3 5 hours This product could be useful for studying angiogenesis regulation and cancer progression