Learn More
Meso Scale Discovery R-PLEX Human Enolase 2 ASY
SDP
Catalog No. 5000080211
Encompass
The R-PLEX Human Enolase 2 Assay Kit from Meso Scale Discovery is used to quantitatively measure enolase 2 in human serum plasma and cell culture supernatants It utilizes an electrochemiluminescent detection method and provides high sensitivity and broad dynamic range The assay takes approximately 4 hours to complete and requires an MSD instrument such as QuickPlex QuickPlex SQ 120 or QuickPlex SQ 240 Each kit provides reagents for one 96-well plate which can assay 40 samples in duplicate
Catalog No. 50-000-80211 Supplier Meso Scale Discovery Supplier No. K1512DR2
The R-PLEX Human Enolase 2 Assay Kit from Meso Scale Discovery is used to quantitatively measure enolase 2 in human serum plasma and cell culture supernatants It utilizes an electrochemiluminescent detection method and provides high sensitivity and broad dynamic range The assay takes approximately 4 hours to complete and requires an MSD instrument such as QuickPlex QuickPlex SQ 120 or QuickPlex SQ 240 Each kit provides reagents for one 96-well plate which can assay 40 samples in duplicate