Learn More
Meso Scale Discovery R-PLEX Human Enolase 2 ASYQP
SDP
Catalog No. 5000080212
Encompass
The R-PLEX Human Enolase 2 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring Enolase 2 protein in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Enolase 2 a glycolytic enzyme in neurons and neuroendocrine tissues may serve as a biomarker for neuronal injury enabling quantitative measurement in biological samples
Catalog No. 50-000-80212 Supplier Meso Scale Discovery Supplier No. K1512DR22
The R-PLEX Human Enolase 2 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring Enolase 2 protein in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Enolase 2 a glycolytic enzyme in neurons and neuroendocrine tissues may serve as a biomarker for neuronal injury enabling quantitative measurement in biological samples