Learn More
Meso Scale Discovery R-PLEX Human Ep-CAM ASY
SDP
Catalog No. 5000079878
Encompass
The R-PLEX Human EP CAM Assay is a multiplex immunoassay panel that measures EPCAM in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and high precision with low sample volume requirements This assay kit includes one plate that can test up to 80 samples in duplicate along with assay diluent detection antibody concentrate read buffer and standards It is suitable for measuring EPCAM levels in biological research and drug discovery applications
Catalog No. 50-000-79878 Supplier Meso Scale Discovery Supplier No. K151ACCR2
The R-PLEX Human EP CAM Assay is a multiplex immunoassay panel that measures EPCAM in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and high precision with low sample volume requirements This assay kit includes one plate that can test up to 80 samples in duplicate along with assay diluent detection antibody concentrate read buffer and standards It is suitable for measuring EPCAM levels in biological research and drug discovery applications