Learn More
Meso Scale Discovery R-PLEX Human Ep-CAM ASYQP
SDP
Catalog No. 5000078859
Encompass
The R-PLEX Human EP CAM Assay is a multiplex immunoassay panel that measures EPCAM in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay allows simultaneous measurement of EPCAM along with other biomarkers It has been validated on the MESO QuickPlex SQ 120 instrument and is suitable for cancer research and biomarker discovery The panel contains a plate coated with capture antibodies detection antibodies calibrators and controls The assay takes 3 5 hours to complete
Catalog No. 50-000-78859 Supplier Meso Scale Discovery Supplier No. K151ACCR22
The R-PLEX Human EP CAM Assay is a multiplex immunoassay panel that measures EPCAM in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay allows simultaneous measurement of EPCAM along with other biomarkers It has been validated on the MESO QuickPlex SQ 120 instrument and is suitable for cancer research and biomarker discovery The panel contains a plate coated with capture antibodies detection antibodies calibrators and controls The assay takes 3 5 hours to complete