Learn More
Meso Scale Discovery R-PLEX Human ER- ASY
SDP
Catalog No. 5000081929
Encompass
The R-PLEX Human ER alpha Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures estrogen receptor alpha (ER ) in human cell lysates and tissue homogenates The assay uses electrochemiluminescence (ECL) detection technology and provides a rapid sensitive and reproducible method to quantify ER levels from as little as 15 g of total protein This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with an ECL tag calibrator diluent wash buffer read buffer and vials of lyophilized calibrator The kit can perform up to 80 tests in duplicate and has a testing time of 2 5 hours
Catalog No. 50-000-81929 Supplier Meso Scale Discovery Supplier No. K151AKPR2
The R-PLEX Human ER alpha Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures estrogen receptor alpha (ER ) in human cell lysates and tissue homogenates The assay uses electrochemiluminescence (ECL) detection technology and provides a rapid sensitive and reproducible method to quantify ER levels from as little as 15 g of total protein This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with an ECL tag calibrator diluent wash buffer read buffer and vials of lyophilized calibrator The kit can perform up to 80 tests in duplicate and has a testing time of 2 5 hours