Learn More
Meso Scale Discovery R-PLEX Human ER- ASYQP
SDP
Catalog No. 5000081628
Encompass
The R-PLEX Human ER alpha Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures estrogen receptor alpha (ER ) in human cell lysates and tissue extracts The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample input The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and reagents required to perform the assay Results are obtained in as little as 3 5 hours This assay allows for the accurate quantification of ER across samples
Catalog No. 50-000-81628 Supplier Meso Scale Discovery Supplier No. K151AKPR22
The R-PLEX Human ER alpha Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures estrogen receptor alpha (ER ) in human cell lysates and tissue extracts The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample input The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and reagents required to perform the assay Results are obtained in as little as 3 5 hours This assay allows for the accurate quantification of ER across samples