Learn More
Meso Scale Discovery R-PLEX Human ERbB2 ASY
SDP
Catalog No. 5000078941
Encompass
The R-PLEX Human ERBB2 Assay is a multiplex immunoassay panel that measures ERBB2 protein levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and is compatible with the SECTOR S 600 instrument The panel allows for the simultaneous quantitative measurement of ERBB2 along with other biomarkers This assay has high sensitivity broad dynamic range and requires low sample volume The R-PLEX Human ERBB2 Assay provides a reproducible and robust tool for cancer research and drug development applications
Catalog No. 50-000-78941 Supplier Meso Scale Discovery Supplier No. K151ACDR2
The R-PLEX Human ERBB2 Assay is a multiplex immunoassay panel that measures ERBB2 protein levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and is compatible with the SECTOR S 600 instrument The panel allows for the simultaneous quantitative measurement of ERBB2 along with other biomarkers This assay has high sensitivity broad dynamic range and requires low sample volume The R-PLEX Human ERBB2 Assay provides a reproducible and robust tool for cancer research and drug development applications