Learn More
Meso Scale Discovery R-PLEX Human ERbB2 ASYQP
SDP
Catalog No. 5000079422
Encompass
The R-PLEX Human ERBB2 Assay is a multiplex immunoassay panel that measures ERBB2 protein levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides accurate reproducible quantitation of ERBB2 levels This assay kit includes a plate coated with capture antibodies detection antibodies calibrators controls wash buffers substrate and consumables needed to run the assay It has a detection range of 50-25 000 pg/mL and the total assay time is approximately 3 5 hours This product enables sensitive high-throughput profiling of ERBB2 expression levels in biological samples
Catalog No. 50-000-79422 Supplier Meso Scale Discovery Supplier No. K151ACDR22
The R-PLEX Human ERBB2 Assay is a multiplex immunoassay panel that measures ERBB2 protein levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides accurate reproducible quantitation of ERBB2 levels This assay kit includes a plate coated with capture antibodies detection antibodies calibrators controls wash buffers substrate and consumables needed to run the assay It has a detection range of 50-25 000 pg/mL and the total assay time is approximately 3 5 hours This product enables sensitive high-throughput profiling of ERBB2 expression levels in biological samples