Learn More
Meso Scale Discovery R-PLEX Human ErbB3/HER3 ASY
SDP
Supplier:  Meso Scale Discovery K151AJKR2
Encompass
The R-PLEX Human ERBB3 (HER3) Assay is a multiplex immunoassay kit by Meso Scale Diagnostics that enables the simultaneous quantitative measurement of ERBB3 and phospho-ERBB3 across multiple sites such as Y1289 Y1197 Y1262 Y1328 Y1199 Y1222 Y1276 and Y1298 It utilizes electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This assay allows the study of ERBB3 signaling in human cell lines and tissues It requires an MSD instrument such as SECTOR Imager 2400 or 6000 for analysis
Catalog No. 50-000-79609