Learn More
Meso Scale Discovery R-PLEX Human ErbB3/HER3 ASYQP
SDP
Catalog No. 5000081786
Encompass
The R-Plex Human ERBB3 (HER3) Assay by Meso Scale Diagnostics is a multiplex immunoassay kit for measuring human ERBB3 and phospho-ERBB3 at sites Y1289 Y1197 Y1276 Y1262 Y1199 and Y1222 Using MSD s SECTOR technology this electrochemiluminescent assay offers a rapid ultra-sensitive method for profiling ERBB3 signaling in cell and tissue lysates It features a broad dynamic range and high-throughput format ideal for biomarker research and drug discovery related to HER3 The kit includes a 96-well plate with capture antibodies detection antibodies and calibrator diluents
Catalog No. 50-000-81786 Supplier Meso Scale Discovery Supplier No. K151AJKR22
The R-Plex Human ERBB3 (HER3) Assay by Meso Scale Diagnostics is a multiplex immunoassay kit for measuring human ERBB3 and phospho-ERBB3 at sites Y1289 Y1197 Y1276 Y1262 Y1199 and Y1222 Using MSD s SECTOR technology this electrochemiluminescent assay offers a rapid ultra-sensitive method for profiling ERBB3 signaling in cell and tissue lysates It features a broad dynamic range and high-throughput format ideal for biomarker research and drug discovery related to HER3 The kit includes a 96-well plate with capture antibodies detection antibodies and calibrator diluents