Learn More
Meso Scale Discovery R-PLEX Human FABP3/H-FABP ASY
SDP
Catalog No. 5000080283
Encompass
The R-PLEX Human FABP3 (H-FABP) Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures human Heart-type Fatty Acid Binding Protein (H-FABP) in serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibody detection antibody Read Buffer Sample Diluent and Calibrator This assay can detect H-FABP levels as low as 19 pg/mL with an approximate 3 5 hour total assay time
Catalog No. 50-000-80283 Supplier Meso Scale Discovery Supplier No. K1514TR2
The R-PLEX Human FABP3 (H-FABP) Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures human Heart-type Fatty Acid Binding Protein (H-FABP) in serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibody detection antibody Read Buffer Sample Diluent and Calibrator This assay can detect H-FABP levels as low as 19 pg/mL with an approximate 3 5 hour total assay time