Learn More
Meso Scale Discovery R-PLEX Human FABP4/A-FABP ASY
SDP
Catalog No. 5000078901
Encompass
The R-PLEX Human FABP4 (A-FABP) Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of Fatty Acid Binding Protein 4 (FABP4) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the rapid reproducible analysis of FABP4 an important biomarker for obesity insulin resistance type 2 diabetes and cardiovascular disease With minimal hands-on time this kit enables the high-throughput profiling of FABP4 levels in research studies
Catalog No. 50-000-78901 Supplier Meso Scale Discovery Supplier No. K151ABMR2
The R-PLEX Human FABP4 (A-FABP) Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of Fatty Acid Binding Protein 4 (FABP4) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the rapid reproducible analysis of FABP4 an important biomarker for obesity insulin resistance type 2 diabetes and cardiovascular disease With minimal hands-on time this kit enables the high-throughput profiling of FABP4 levels in research studies