Learn More
Meso Scale Discovery R-PLEX Human FasL ASY
SDP
Catalog No. 5000080095
Encompass
The R-PLEX Human FasL Assay Kit from Meso Scale Discovery measures human Fas ligand (FasL) in various biological samples using a multiplex immunoassay This electrochemiluminescence (ECL) assay employs MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture and detection antibodies calibrators controls buffers and seals It offers a simple workflow with no wash steps and results are analyzed on MSD s systems enabling accurate FasL quantitation for apoptosis research
Catalog No. 50-000-80095 Supplier Meso Scale Discovery Supplier No. K151ZBR2
The R-PLEX Human FasL Assay Kit from Meso Scale Discovery measures human Fas ligand (FasL) in various biological samples using a multiplex immunoassay This electrochemiluminescence (ECL) assay employs MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture and detection antibodies calibrators controls buffers and seals It offers a simple workflow with no wash steps and results are analyzed on MSD s systems enabling accurate FasL quantitation for apoptosis research