Learn More
Meso Scale Discovery R-PLEX Human FasL ASYQP
SDP
Catalog No. 5000079217
Encompass
The R-PLEX Human FasL Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human Fas ligand (FasL) in cell culture supernatants serum plasma and other biological samples This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized calibrator and controls to enable the quantification of human FasL This product can aid in the study of apoptosis and immune system function
Catalog No. 50-000-79217 Supplier Meso Scale Discovery Supplier No. K151ZBR22
The R-PLEX Human FasL Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human Fas ligand (FasL) in cell culture supernatants serum plasma and other biological samples This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized calibrator and controls to enable the quantification of human FasL This product can aid in the study of apoptosis and immune system function