Learn More
Meso Scale Discovery R-PLEX Human Ferritin ASY
SDP
Catalog No. 5000079507
Encompass
The R-PLEX Human Ferritin Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of ferritin in human serum plasma and other biological fluids This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a highly sensitive and reproducible method for detecting ferritin levels between 0 53-1000 ng/mL The assay requires a small sample volume of 25 L and can be read on MSD s QuickPlex SQ 120 instrument The R-PLEX Ferritin Assay provides a rapid accurate way to assess iron stores and aid in the diagnosis of iron deficiency or overload
Catalog No. 50-000-79507 Supplier Meso Scale Discovery Supplier No. K151ADAR2
The R-PLEX Human Ferritin Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of ferritin in human serum plasma and other biological fluids This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a highly sensitive and reproducible method for detecting ferritin levels between 0 53-1000 ng/mL The assay requires a small sample volume of 25 L and can be read on MSD s QuickPlex SQ 120 instrument The R-PLEX Ferritin Assay provides a rapid accurate way to assess iron stores and aid in the diagnosis of iron deficiency or overload