Learn More
Meso Scale Discovery R-PLEX Human Ferritin ASYQP
SDP
Catalog No. 5000078946
Encompass
The R-PLEX Human Ferritin Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of ferritin in human serum plasma and other biological fluids This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a highly sensitive and reproducible method for detecting ferritin levels between 0 53-2000 ng/mL The assay requires a small sample volume of 25 L and can be read on MSD s QuickPlex SQ 120 instrument The R-PLEX Ferritin Assay provides a simple rapid and robust tool for monitoring body iron stores and iron deficiency
Catalog No. 50-000-78946 Supplier Meso Scale Discovery Supplier No. K151ADAR22
The R-PLEX Human Ferritin Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of ferritin in human serum plasma and other biological fluids This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide a highly sensitive and reproducible method for detecting ferritin levels between 0 53-2000 ng/mL The assay requires a small sample volume of 25 L and can be read on MSD s QuickPlex SQ 120 instrument The R-PLEX Ferritin Assay provides a simple rapid and robust tool for monitoring body iron stores and iron deficiency