Learn More
Meso Scale Discovery R-PLEX Human FGF-19 ASYQP
SDP
Catalog No. 5000080389
Encompass
The Meso Scale Discovery R-PLEX Human FGF-19 Assay Kit is a multiplex immunoassay that quantitatively measures human FGF-19 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator that must be reconstituted before use The assay can detect FGF-19 concentrations from 1 22 pg/mL to 50 000 pg/mL
Catalog No. 50-000-80389 Supplier Meso Scale Discovery Supplier No. K151AARR22
The Meso Scale Discovery R-PLEX Human FGF-19 Assay Kit is a multiplex immunoassay that quantitatively measures human FGF-19 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator that must be reconstituted before use The assay can detect FGF-19 concentrations from 1 22 pg/mL to 50 000 pg/mL