Learn More
Meso Scale Discovery R-PLEX Human FGF-7 ASYQP
SDP
Catalog No. 5000079127
Encompass
The R-PLEX Human FGF-7 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures FGF-7 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It requires 50 L of sample per well and can assay 46 samples in duplicate The total assay time is 3 5 hours
Catalog No. 50-000-79127 Supplier Meso Scale Discovery Supplier No. K151N4R22
The R-PLEX Human FGF-7 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures FGF-7 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It requires 50 L of sample per well and can assay 46 samples in duplicate The total assay time is 3 5 hours