Learn More
Meso Scale Discovery R-PLEX Human FGF acidic ASY
SDP
Catalog No. 5000081960
Encompass
The R-PLEX Human FGF Acidic Assay Kit from Meso Scale Discovery is used to quantitatively measure human FGF acidic (FGF-1) in serum plasma cell culture supernatants cell and tissue lysates and other biological samples It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity and shows no cross-reactivity with other proteins It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrators and controls The kit can perform up to 94 tests in duplicate
Catalog No. 50-000-81960 Supplier Meso Scale Discovery Supplier No. K151W0R2
The R-PLEX Human FGF Acidic Assay Kit from Meso Scale Discovery is used to quantitatively measure human FGF acidic (FGF-1) in serum plasma cell culture supernatants cell and tissue lysates and other biological samples It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity and shows no cross-reactivity with other proteins It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrators and controls The kit can perform up to 94 tests in duplicate