Learn More
Meso Scale Discovery R-PLEX Human FGF-BP1 ASY
SDP
Catalog No. 5000079452
Encompass
The R-PLEX Human FGF BP1 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the simultaneous quantitative measurement of human FGF Basic Protein 1 in cell and tissue lysates serum plasma (EDTA) saliva and urine It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators controls 10X diluent and wash buffer It can accurately quantify FGF BP1 levels between 0 61 - 10 000 pg/mL with an incubation time of 2 hours
Catalog No. 50-000-79452 Supplier Meso Scale Discovery Supplier No. K151ABNR2
The R-PLEX Human FGF BP1 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the simultaneous quantitative measurement of human FGF Basic Protein 1 in cell and tissue lysates serum plasma (EDTA) saliva and urine It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators controls 10X diluent and wash buffer It can accurately quantify FGF BP1 levels between 0 61 - 10 000 pg/mL with an incubation time of 2 hours