Learn More
Meso Scale Discovery R-PLEX Human FGF-BP1 ASYQP
SDP
Catalog No. 5000079573
Encompass
The R-PLEX Human FGF BP1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of FGF BP1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid time-to-result This assay requires low sample volume and has minimal cross-reactivity It can be used for biomarker research disease monitoring and drug discovery and development The kit includes all necessary reagents and standards for analyzing 1 x 96-well plate
Catalog No. 50-000-79573 Supplier Meso Scale Discovery Supplier No. K151ABNR22
The R-PLEX Human FGF BP1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of FGF BP1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid time-to-result This assay requires low sample volume and has minimal cross-reactivity It can be used for biomarker research disease monitoring and drug discovery and development The kit includes all necessary reagents and standards for analyzing 1 x 96-well plate