Learn More
Meso Scale Discovery R-PLEX Human GAD2/GAD-65 ASY
SDP
Catalog No. 5000080496
Encompass
The Meso Scale Discovery GAD2-GAD-65 kit is a multiplex immunoassay that measures GAD65 and GAD67 autoantibodies in human serum and plasma It uses electrochemiluminescence detection technology and provides quantitative results in under 5 hours This assay kit contains all necessary reagents and plates for analyzing autoantibodies implicated in type 1 diabetes and other autoimmune conditions With high sensitivity and specificity it enables the differential detection of GAD65 and GAD67 autoantibodies which can aid in disease diagnosis and progression monitoring The kit has a testing capacity of 96 wells and shows minimal cross-reactivity and interference
Catalog No. 50-000-80496 Supplier Meso Scale Discovery Supplier No. K151AKBR2
The Meso Scale Discovery GAD2-GAD-65 kit is a multiplex immunoassay that measures GAD65 and GAD67 autoantibodies in human serum and plasma It uses electrochemiluminescence detection technology and provides quantitative results in under 5 hours This assay kit contains all necessary reagents and plates for analyzing autoantibodies implicated in type 1 diabetes and other autoimmune conditions With high sensitivity and specificity it enables the differential detection of GAD65 and GAD67 autoantibodies which can aid in disease diagnosis and progression monitoring The kit has a testing capacity of 96 wells and shows minimal cross-reactivity and interference