Learn More
Meso Scale Discovery R-PLEX Human GAD2/GAD-65 ASYQP
SDP
Catalog No. 5000080497
Encompass
The R-Plex Human GAD2 (GAD 65) Antibody Set from Meso Scale Discovery is used to measure glutamic acid decarboxylase 2 (GAD2) in human serum plasma tissue lysates or cell culture supernatants using an electrochemiluminescent multiplex immunoassay on Meso Scale Discovery instrumentation This set contains a capture antibody and a detection antibody for GAD2 allowing sensitive and specific quantification of this protein GAD2 is an enzyme involved in synthesizing the neurotransmitter GABA and has been associated with neurological disorders This set can aid in biomarker research related to GAD2
Catalog No. 50-000-80497 Supplier Meso Scale Discovery Supplier No. K151AKBR22
The R-Plex Human GAD2 (GAD 65) Antibody Set from Meso Scale Discovery is used to measure glutamic acid decarboxylase 2 (GAD2) in human serum plasma tissue lysates or cell culture supernatants using an electrochemiluminescent multiplex immunoassay on Meso Scale Discovery instrumentation This set contains a capture antibody and a detection antibody for GAD2 allowing sensitive and specific quantification of this protein GAD2 is an enzyme involved in synthesizing the neurotransmitter GABA and has been associated with neurological disorders This set can aid in biomarker research related to GAD2