Learn More
Meso Scale Discovery R-PLEX Human Galectin-1 ASY
SDP
Catalog No. 5000082309
Encompass
The Meso Scale Discovery R-PLEX Human Galectin-1 Assay Kit is a multiplex immunoassay that quantitatively measures galectin-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions for generating standard curves and assessing assay performance The kit enables reliable quantification of galectin-1 levels from small sample volumes
Catalog No. 50-000-82309 Supplier Meso Scale Discovery Supplier No. K151Q8R2
The Meso Scale Discovery R-PLEX Human Galectin-1 Assay Kit is a multiplex immunoassay that quantitatively measures galectin-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions for generating standard curves and assessing assay performance The kit enables reliable quantification of galectin-1 levels from small sample volumes