Learn More
Meso Scale Discovery R-PLEX Human Galectin-1 ASYQP
SDP
Catalog No. 5000082190
Encompass
The R-PLEX Human Galectin-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures human Galectin-1 in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator The assay requires 2 5 hours to complete can detect Galectin-1 concentrations between 1 53 - 1000 pg/mL and is suitable for analyzing 50 L samples
Catalog No. 50-000-82190 Supplier Meso Scale Discovery Supplier No. K151Q8R22
The R-PLEX Human Galectin-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures human Galectin-1 in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator The assay requires 2 5 hours to complete can detect Galectin-1 concentrations between 1 53 - 1000 pg/mL and is suitable for analyzing 50 L samples