Learn More
Meso Scale Discovery R-PLEX Human GAS-6/AXL-L ASY
SDP
Catalog No. 5000082087
Encompass
The R-PLEX Human Gas6/Axl-L Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human Gas6 and Axl proteins in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents It quantifies Gas6 and Axl crucial for cell survival proliferation migration and phagocytosis in 3 5 hours performing 39 tests in duplicate
Catalog No. 50-000-82087 Supplier Meso Scale Discovery Supplier No. K151G5R2
The R-PLEX Human Gas6/Axl-L Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human Gas6 and Axl proteins in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents It quantifies Gas6 and Axl crucial for cell survival proliferation migration and phagocytosis in 3 5 hours performing 39 tests in duplicate