Learn More
Meso Scale Discovery R-PLEX Human GCPII/PSMA ASY
SDP
Catalog No. 5000078222
Encompass
The R-PLEX Human GCPII (PSMA) Assay is a multiplex immunoassay panel that measures GCPII (PSMA) in human serum plasma and cell culture supernatants It utilizes Meso Scale Discovery electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay measures total GCPII and includes reagents for 100 tests in one sector (5-plex) of an R-PLEX plate GCPII also known as PSMA is highly expressed in prostate tissues and can serve as a biomarker for prostate cancer progression This assay allows sensitive and reproducible quantification of GCPII levels
Catalog No. 50-000-78222 Supplier Meso Scale Discovery Supplier No. K151ABPR2
The R-PLEX Human GCPII (PSMA) Assay is a multiplex immunoassay panel that measures GCPII (PSMA) in human serum plasma and cell culture supernatants It utilizes Meso Scale Discovery electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay measures total GCPII and includes reagents for 100 tests in one sector (5-plex) of an R-PLEX plate GCPII also known as PSMA is highly expressed in prostate tissues and can serve as a biomarker for prostate cancer progression This assay allows sensitive and reproducible quantification of GCPII levels