Learn More
Meso Scale Discovery R-PLEX Human GCPII/PSMA ASYQP
SDP
Catalog No. 5000079876
Encompass
The R-PLEX Human GCPII (PSMA) Assay Sector 5 PL Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures GCPII (PSMA) levels in human serum plasma and cell culture supernatant samples This assay kit utilizes electrochemiluminescence (ECL) detection technology and provides reagents for analyzing 3 plates with 96 wells per plate The assay has a testing range of 0 064 - 400 ng/mL and exhibits high sensitivity and specificity This product requires the MSD QuickPlex SQ120 instrument and provides all necessary reagents and plates for quantitative analysis of GCPII (PSMA)
Catalog No. 50-000-79876 Supplier Meso Scale Discovery Supplier No. K151ABPR22
The R-PLEX Human GCPII (PSMA) Assay Sector 5 PL Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures GCPII (PSMA) levels in human serum plasma and cell culture supernatant samples This assay kit utilizes electrochemiluminescence (ECL) detection technology and provides reagents for analyzing 3 plates with 96 wells per plate The assay has a testing range of 0 064 - 400 ng/mL and exhibits high sensitivity and specificity This product requires the MSD QuickPlex SQ120 instrument and provides all necessary reagents and plates for quantitative analysis of GCPII (PSMA)