Learn More
Meso Scale Discovery R-PLEX Human GDF-15 Assay, Singleplex, 5PL
SDP
Catalog No. NC2457984
Encompass
The MESO SCALE DISCOVERY R-PLEX Human GDF-15 Assay is a singleplex immunoassay designed for the quantitative measurement of Human GDF-15. This assay utilizes the R-PLEX platform, providing essential components for constructing a reliable assay for research purposes. Its flexible system allows for potential integration into multiplex panels with other R-PLEX analytes. The assay is intended only for research use and includes comprehensive documentation such as a product insert and datasheet.
  • Target analyte is Human GDF-15
  • Singleplex assay format
  • Utilizes R-PLEX platform
  • Research use only designation
  • Multiplexing capability with other R-PLEX analytes
  • Essential components provided for assay development
  • Documentation available includes product insert and datasheet
Catalog No. NC2457984 Supplier Meso Scale Discovery Supplier No. K151YDR2
The MESO SCALE DISCOVERY R-PLEX Human GDF-15 Assay is a singleplex immunoassay designed for the quantitative measurement of Human GDF-15. This assay utilizes the R-PLEX platform, providing essential components for constructing a reliable assay for research purposes. Its flexible system allows for potential integration into multiplex panels with other R-PLEX analytes. The assay is intended only for research use and includes comprehensive documentation such as a product insert and datasheet.
  • Target analyte is Human GDF-15
  • Singleplex assay format
  • Utilizes R-PLEX platform
  • Research use only designation
  • Multiplexing capability with other R-PLEX analytes
  • Essential components provided for assay development
  • Documentation available includes product insert and datasheet