Learn More
Meso Scale Discovery R-PLEX Human GDF-15 ASYQP
SDP
Catalog No. 5000078783
Encompass
The Meso Scale Discovery R-PLEX Human GDF-15 Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of GDF-15 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms The GDF-15 assay measures a range of 50-100 000 pg/mL with minimal cross-reactivity to related analytes It is intended for research use only to aid in understanding GDF-15 biology and disease processes
Catalog No. 50-000-78783 Supplier Meso Scale Discovery Supplier No. K151YDR22
The Meso Scale Discovery R-PLEX Human GDF-15 Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of GDF-15 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms The GDF-15 assay measures a range of 50-100 000 pg/mL with minimal cross-reactivity to related analytes It is intended for research use only to aid in understanding GDF-15 biology and disease processes