Learn More
Meso Scale Discovery R-PLEX Human GDF-8 ASY
SDP
Catalog No. 5000079461
Encompass
The R-PLEX Human GDF-8 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of GDF-8 (also known as myostatin) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be run on Meso Scale Diagnostic s SECTOR Imagers and QuickPlex plate readers It includes a plate coated with capture antibodies detection antibodies calibrators and reagents required for the assay
Catalog No. 50-000-79461 Supplier Meso Scale Discovery Supplier No. K151ACER2
The R-PLEX Human GDF-8 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the quantitative measurement of GDF-8 (also known as myostatin) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be run on Meso Scale Diagnostic s SECTOR Imagers and QuickPlex plate readers It includes a plate coated with capture antibodies detection antibodies calibrators and reagents required for the assay