Learn More
Meso Scale Discovery R-PLEX Human GDF-8 ASYQP
SDP
Catalog No. 5000078914
Encompass
The R-PLEX Human GDF-8 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics for measuring GDF-8 (myostatin) in human serum plasma and cell culture supernatant It uses electrochemiluminescence (ECL) detection offering high sensitivity a broad dynamic range and low sample volume requirements Compatible with SECTOR Imager platforms data is analyzed via MSD Discovery Workbench software The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation and assay validation
Catalog No. 50-000-78914 Supplier Meso Scale Discovery Supplier No. K151ACER22
The R-PLEX Human GDF-8 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics for measuring GDF-8 (myostatin) in human serum plasma and cell culture supernatant It uses electrochemiluminescence (ECL) detection offering high sensitivity a broad dynamic range and low sample volume requirements Compatible with SECTOR Imager platforms data is analyzed via MSD Discovery Workbench software The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation and assay validation