Learn More
Meso Scale Discovery R-PLEX Human GFAP ASY
SDP
Catalog No. 5000080206
Encompass
The R-PLEX Human GFAP Assay Kit from Meso Scale Discovery measures glial fibrillary acidic protein (GFAP) in human samples using electrochemiluminescence (ECL) technology It offers high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture and detection antibodies Read Buffer Sample Diluent and calibrators for standard curve generation It quantifies GFAP a biomarker in astrocytes linked to CNS disorders with a 2 5-hour protocol for high-throughput profiling
Catalog No. 50-000-80206 Supplier Meso Scale Discovery Supplier No. K1511MR2
The R-PLEX Human GFAP Assay Kit from Meso Scale Discovery measures glial fibrillary acidic protein (GFAP) in human samples using electrochemiluminescence (ECL) technology It offers high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture and detection antibodies Read Buffer Sample Diluent and calibrators for standard curve generation It quantifies GFAP a biomarker in astrocytes linked to CNS disorders with a 2 5-hour protocol for high-throughput profiling