Learn More
Meso Scale Discovery R-PLEX Human GRP ASY
SDP
Catalog No. 5000080154
Encompass
The Meso Scale Discovery GRP Human Proinflammatory Panel 1 Kit is a multiplex immunoassay that simultaneously measures Granulocyte-Macrophage Colony-Stimulating Factor (GM-CSF) Interferon gamma (IFN-) Interleukin-1 beta (IL-1 ) Interleukin-2 (IL-2) Interleukin-4 (IL-4) Interleukin-6 (IL-6) Interleukin-8 (IL-8) Interleukin-10 (IL-10) Interleukin-12p70 (IL-12p70) Interleukin-13 (IL-13) and Tumor Necrosis Factor alpha (TNF- ) in serum plasma and cell culture supernatants using electrochemiluminescence detection technology This 11-plex panel allows high throughput sensitive and reproducible quantification of multiple inflammatory biomarkers from a small sample volume
Catalog No. 50-000-80154 Supplier Meso Scale Discovery Supplier No. K151ALCR2
The Meso Scale Discovery GRP Human Proinflammatory Panel 1 Kit is a multiplex immunoassay that simultaneously measures Granulocyte-Macrophage Colony-Stimulating Factor (GM-CSF) Interferon gamma (IFN-) Interleukin-1 beta (IL-1 ) Interleukin-2 (IL-2) Interleukin-4 (IL-4) Interleukin-6 (IL-6) Interleukin-8 (IL-8) Interleukin-10 (IL-10) Interleukin-12p70 (IL-12p70) Interleukin-13 (IL-13) and Tumor Necrosis Factor alpha (TNF- ) in serum plasma and cell culture supernatants using electrochemiluminescence detection technology This 11-plex panel allows high throughput sensitive and reproducible quantification of multiple inflammatory biomarkers from a small sample volume