Learn More
Meso Scale Discovery R-PLEX Human GRP ASYQP
SDP
Catalog No. 5000078434
Encompass
The K151ALCR-22 is a 22-pin DIP socket from Mesoscale It has a 0 05 (1 27mm) pitch and accepts 0 025 (0 635mm) square pins The socket is made from glass reinforced UL94 V-0 rated liquid crystal polymer with gold plated beryllium copper alloy contacts It can operate from -55 C to 125 C The socket has a current rating of 1A per pin It is RoHS compliant
Catalog No. 50-000-78434 Supplier Meso Scale Discovery Supplier No. K151ALCR22
The K151ALCR-22 is a 22-pin DIP socket from Mesoscale It has a 0 05 (1 27mm) pitch and accepts 0 025 (0 635mm) square pins The socket is made from glass reinforced UL94 V-0 rated liquid crystal polymer with gold plated beryllium copper alloy contacts It can operate from -55 C to 125 C The socket has a current rating of 1A per pin It is RoHS compliant